Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ERK2 Monoclonal Antibody (A10D2), Huabio

Catalog No. 502544800
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-4800 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-4800 Supplier HUABIO RESEARCH INC Supplier No. EM190154
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ERK2 Monoclonal antibody specifically detects ERK2 in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen ERK2
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone A10D2
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene MAPK1
Gene Accession No. P28482, P63085
Gene Alias 9030612K14Rik; AA407128; AU018647; C78273; ERK; ERK2; ERK-2; ERT1; extracellular signal-regulated kinase 2; extracellular signal-regulated kinase-2; extracellular-signal-regulated kinase 2; MAP kinase 1; MAP kinase 2; MAP kinase isoform p42; Mapk; MAPK 1; MAPK 2; Mapk1; MAPK2; mitogen activated protein kinase 1; mitogen-activated protein kinase 1; Mitogen-activated protein kinase 2; p38; p40; p41; p41mapk; P42MAPK; p42-MAPK; Prkm1; PRKM2; protein tyrosine kinase ERK2
Host Species Mouse
Immunogen Recombinant protein within human ERK2 aa 200-360.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26413, 5594
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.