Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ERK2Rabbit anti-Human, Polyclonal, Rockland™
SDP

Catalog No. 501990043
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-0043 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-199-0043 Supplier Rockland Immunochemicals Supplier No. 100401218
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This antiserum is directed against human p42 MAP Kinase (ERK2) protein and is useful in determining its presence by immunoblotting. No reactivity is observed against p44 MAP Kinase (ERK1). Cross reactivity is expected with p44 MAP Kinase (ERK1) proteins from human and mouse sources. Reactivity to rat tissues is also anticipated due to high sequence homology. Reactivity against homologues from other sources is not known.

ERK2 (also known as MAPK1) is a member of the MAP kinase subfamily of Ser/Thr protein kinases. MAP kinases, also known as extracellular signal-regulated kinases (ERKs), act as an integration point for multiple biochemical signals, and are involved in a wide variety of cellular processes such as proliferation, differentiation, transcription regulation and development. The activation of this kinase requires its phosphorylation by upstream kinases. Upon activation, this kinase translocates to the nucleus of the stimulated cells, where it phosphorylates nuclear targets.

Specifications

Antigen ERK2
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 85 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene MAPK1
Gene Accession No. P28482
Gene Alias 9030612K14Rik; AA407128; AU018647; C78273; ERK; Erk2; ERK-2; ERT1; Extracellular signal-regulated kinase 2; extracellular signal-regulated kinase-2; extracellular-signal-regulated kinase 2; MAP kinase 1; MAP kinase 2; MAP kinase isoform p42; Mapk; MAPK 1; MAPK 2; MAPK1; MAPK2; mitogen activated protein kinase 1; Mitogen-activated protein kinase 1; Mitogen-activated protein kinase 2; p38; p40; p41; p41mapk; p42mapk; p42-MAPK; PRKM1; PRKM2; protein tyrosine kinase ERK2
Gene Symbols MAPK1
Host Species Rabbit
Immunogen Anti-Erk2 antibody was prepared by repeated immunizations with an Erk2 containing fusion protein. The epitope maps near the carboxy-terminus of human p42 MAP Kinase (ERK2) protein. The epitope is identical to the corresponding sequence in mouse and differs from the rat sequence by a single, conservative amino acid substitution.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5594
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.