Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ERK3/MAPK6 Antibody (OTI5E1), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271242F
Description
ERK3/MAPK6 Monoclonal specifically detects ERK3/MAPK6 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
ERK3/MAPK6 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
MAPK6 | |
Recombinant protein expressed in E.coli corresponding to amino acids 345-721 of human ERK3/MAPK6. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG2a |
Immunocytochemistry, Immunofluorescence | |
OTI5E1 | |
Immunocytochemistry/Immunofluorescence | |
DKFZp686F03189, EC 2.7.11, EC 2.7.11.24, ERK-3, ERK3p97-MAPK, Extracellular signal-regulated kinase 3, extracellular signal-regulated kinase, p97, HsT17250, MAP kinase 6, MAP kinase isoform p97, MAPK 6, mitogen-activated protein kinase 6, p97MAPK, PRKM6, protein kinase, mitogen-activated 5, protein kinase, mitogen-activated 6 | |
Mouse | |
Affinity Purified | |
Breast Cancer, Cancer, Cell Cycle and Replication, Cellular Markers, Hypoxia, Phospho Specific, Signal Transduction, Stem Cell Markers | |
5597.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction