Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ERK3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583134
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583134 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583134 Supplier Invitrogen™ Supplier No. PA583134
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YSFPMDEPIS SHPFHIEDEV DDILLMDETH SHIYNWERYH DCQFSEHDWP VHNNFDIDEV QLDPRALSDV TDEEEVQVDP RKYLDGDREK YLEDPAFDTN YSTEPCWQYS DHHENKYCDL ECSH Highest antigen sequence indentity to the following orthologs: Rat - 96%, Mouse - 96%.

The protein encoded by this gene is a member of the Ser/Thr protein kinase family, and is most closely related to mitogen-activated protein kinases (MAP kinases). MAP kinases also known as extracellular signal-regulated kinases (ERKs), are activated through protein phosphorylation cascades and act as integration points for multiple biochemical signals. This kinase is localized in the nucleus, and has been reported to be activated in fibroblasts upon treatment with serum or phorbol esters. Tissue specificity: Highest expression in the skeletal muscle, followed by the brain. Also found in heart, placenta, lung, liver, pancreas, kidney and skin fibroblasts.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ERK3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MAPK6
Gene Accession No. Q16659
Gene Alias 2610021I23Rik; D130053K17Rik; DKFZp686F03189; erk3; ERK-3; Erk-3 related; Extracellular signal-regulated kinase 3; extracellular signal-regulated kinase, p97; HsT17250; LOC538094 protein; MAP kinase 6; MAP kinase isoform p97; MAPK 6; Mapk4; Mapk6; Mapk63; mitogen activated protein kinase 4; mitogen activated protein kinase 6; mitogen-activated protein kinase 6; p55-MAPK; p97mapk; p97-MAPK; Prkm4; Prkm6; protein kinase, mitogen activated kinase 4; protein kinase, mitogen-activated 5; protein kinase, mitogen-activated 6; si:ch211-235f12.1
Gene Symbols MAPK6
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MAPK6. Recombinant protein control fragment (Product #RP-91423).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5597
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.