Learn More
ERK3 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302654AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Orangutan
ERK3 is a member of the Ser/Thr protein kinase family, and is most closely related to mitogen-activated protein kinases (MAP kinases). MAP kinases also known as extracellular signal-regulated kinases (ERKs), are activated through protein phosphorylation cascades and act as integration points for multiple biochemical signals. This kinase is localized in the nucleus, and has been reported to be activated in fibroblasts upon treatment with serum or phorbol esters.Specifications
ERK3 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q16659, Q61532 | |
MAPK6 | |
Between 671 and 721 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
MAPK6 | |
ERK3, HsT17250, PRKM6, p97MAPK, ERK-3; MAP kinase 6; MAP kinase isoform p97; MAPK 6; extracellular signal-regulated kinase 3; extracellular signal-regulated kinase, p97; p97-MAPK; protein kinase, mitogen-activated 5; protein kinase, mitogen-activated 6 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
50772, 5597 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.