Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ERK4/MAPK4 Antibody (OTI2D7), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271240R
Description
ERK4/MAPK4 Monoclonal specifically detects ERK4/MAPK4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
ERK4/MAPK4 | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
MAPK4 | |
Full length recombinant protein of human ERK4/MAPK4 (NP_002738) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG2b |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI2D7 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
EC 2.7.11, EC 2.7.11.24, ERK3, Erk3-related, Erk4, ERK-4, Extracellular signal-regulated kinase 4, MAP kinase 4, MAP kinase isoform p63, MAPK 4, mitogen-activated protein kinase 4, p63MAPK, PRKM4p63-MAPK | |
Mouse | |
Affinity Purified | |
Cell Cycle and Replication | |
5596.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction