Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ ERK5-IN-1
Click to view available options
Quantity:
10 mg
50 mg
Description
ERK5-IN-1 is a potent and selective ERK5 inhibitor; inhibits EGFR-induced ERK5 autophosphorylation (EC50 = 90 nM) and ERK5 enzymatic activity (IC50 = 162 nM). Exhibits at least 30-fold selectivity for ERK5 over LRRK2 in a cell based assay. Also selective for ERK5 over a panel of other kinases. Orally bioavailable.
Specifications
Specifications
| Purity | >98% |
| Description | ERK5-IN-1 |
| Molecular Weight (g/mol) | 638.8 |
| Product Type | ERK5-IN-1 |
| Quantity | 10 mg |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction