Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ERLIN1 Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NBP145206
Description
ERLIN1 Polyclonal specifically detects ERLIN1 in Human samples. It is validated for Western Blot, Peptide ELISA.Specifications
ERLIN1 | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
Band_7 23-211 Keo4 (Interim) similar to C.elegans protein C42C1.9, C10orf69, chromosome 10 open reading frame 69, Endoplasmic reticulum lipid raft-associated protein 1, ER lipid raft associated 1, Erlin-1, KE04SPFH domain-containing protein 1, KEO4, Protein KE04, SPFH domain family, member 1, SPFH1, Stomatin-prohibitin-flotillin-HflC/K domain-containing protein 1 | |
Goat | |
Affinity Purified | |
RUO | |
10613 | |
Human | |
IgG |
Western Blot, ELISA | |
0.5 mg/mL | |
Western Blot 0.3 - 1 ug/ml, Peptide ELISA Detection limit 1:8000 | |
NP_006450.2 | |
ERLIN1 | |
Peptide with sequence C-EALEPSGENVIQNKE corresponding to C-Terminus according to NP_006450.2. | |
0.1 mg | |
Primary | |
Reported variants represent identical protein: NP_006450.2, NP_001094096.1 | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction