Learn More
ERLIN1 Mouse anti-Human, Clone: OTI4B1, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF809148

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Component of the ERLIN1/ERLIN2 complex which mediates the endoplasmic reticulum-associated degradation (ERAD) of inositol 1,4,5-trisphosphate receptors (IP3Rs). Involved in regulation of cellular cholesterol homeostasis by regulation the SREBP signaling pathway. Binds cholesterol and may promote ER retention of the SCAP-SREBF complex.Specifications
ERLIN1 | |
Monoclonal | |
Unconjugated | |
ERLIN1 | |
C10orf69, Erlin-1, KE04, KEO4, SPFH1, SPG62 | |
Mouse | |
Affinity Chromatography | |
RUO | |
10613 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI4B1 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
O75477 | |
ERLIN1 | |
Human recombit protein fragment corresponding to amino acids 107-346 of human ERLIN1 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.