Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ERLIN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590076
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590076 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590076 Supplier Invitrogen™ Supplier No. PA590076
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KADAEYYAAH KYATSNKHKL TPEYLELKKY QAIASNSKIY FGSNIPNMFV DSSCALKYSD IRTGRESSLP SKEALEPSGE NVIQNKESTG; Positive Samples: 293T, HepG2, BxPC3, DU145, OVCAR-3, Mouse kidney, Rat testis; Cellular Location: Endoplasmic reticulum membrane, Single-pass type II membrane protein.

Component of the ERLIN1/ERLIN2 complex which mediates the endoplasmic reticulum-associated degradation (ERAD) of inositol 1,4,5-trisphosphate receptors (IP3Rs). Involved in regulation of cellular cholesterol homeostasis by regulation the SREBP signaling pathway. Binds cholesterol and may promote ER retention of the SCAP-SREBF complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ERLIN1
Applications ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1.26 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Erlin1
Gene Accession No. O75477, Q91X78
Gene Alias 2810439N09Rik; Band_7 23-211 Keo4 (Interim) similar to C.elegans protein C42C1.9; C10orf69; C80197; endoplasmic reticulum lipid raft-associated protein 1; ER lipid raft associated 1; Erlin1; erlin-1; KE04; Keo4; OTTHUMP00000059425; OTTHUMP00000197867; Protein KE04; Protein KE04 homolog; RGD1307058; SPFH domain family, member 1; SPFH domain-containing protein 1; Spfh1; SPG62; stomatin-prohibitin-flotillin-HflC/K domain-containing protein 1
Gene Symbols Erlin1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 259-348 of human ERLIN1 (NP_0064502).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10613, 226144, 293939
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.