Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ERLIN1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA597715
Description
ERLIN1 Polyclonal Antibody for Western Blot, IHC (P), ELISA
Component of the ERLIN1/ERLIN2 complex which mediates the endoplasmic reticulum-associated degradation (ERAD) of inositol 1,4,5-trisphosphate receptors (IP3Rs). Involved in regulation of cellular cholesterol homeostasis by regulation the SREBP signaling pathway. Binds cholesterol and may promote ER retention of the SCAP-SREBF complex.
Specifications
ERLIN1 | |
Polyclonal | |
Unconjugated | |
Erlin1 | |
2810439N09Rik; Band_7 23-211 Keo4 (Interim) similar to C.elegans protein C42C1.9; C10orf69; C80197; endoplasmic reticulum lipid raft-associated protein 1; ER lipid raft associated 1; Erlin1; erlin-1; KE04; Keo4; OTTHUMP00000059425; OTTHUMP00000197867; Protein KE04; Protein KE04 homolog; RGD1307058; SPFH domain family, member 1; SPFH domain-containing protein 1; Spfh1; SPG62; stomatin-prohibitin-flotillin-HflC/K domain-containing protein 1 | |
Rabbit | |
Protein G | |
RUO | |
10613, 226144 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
4 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
O75477, Q91X78 | |
Erlin1 | |
Recombinant Human Erlin-1 protein (27-346AA). | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction