Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ERLIN2 Monoclonal Antibody (3H9A2)
GREENER_CHOICE

Catalog No. PIMA549296
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA549296 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA549296 Supplier Invitrogen™ Supplier No. MA549296
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human HepG2 whole cell, human T47D whole cell, human HCCT tissue, human HCCP tissue. IHC: human ovarian serous carcinoma tissue, human placenta tissue, human squamous cell lung carcinoma tissue, human thyroiditis tissue. ICC/IF: T47D cell. Flow: JK cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

ERLIN2, also named as C8orf2 and SPFH2, is a component of the ERLIN1/ERLIN2 complex which mediates the endoplasmic reticulum-associated degradation (ERAD) of inositol 1, 4, 5-trisphosphate receptors (IP3Rs) such as ITPR1. It promotes sterol-accelerated ERAD of HMGCR probably implicating an AMFR/gp78-containing ubiquitin ligase complex. ERLIN2 is involved in regulation of cellular cholesterol homeostasis by regulation the SREBP signaling pathway. The MW of ERLIN1/ERLIN2 complex is about 70 kDa.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ERLIN2
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 3H9A2
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and no preservative
Gene ERLIN2
Gene Accession No. O94905
Gene Alias BC036333; C87251; C8orf2; EMBL:AAI68668.1, ECO:0000312; EMBL:AAI68668.1}; endoplasmic reticulum lipid raft-associated protein 2; endoplasmic reticulum lipid raft-associated protein 2 {ECO:0000250; ER lipid raft associated 2; Erlin2; erlin-2; erlin-2 {ECO:0000250; erlin2 {ECO:0000312; HGNC:1356; MGC87072; NET32; RGD:1309010}; RGD1309010; SPFH domain family, member 2; SPFH domain-containing protein 2; SPFH domain-containing protein 2 {ECO:0000250; Spfh2; SPG18; stomatin-prohibitin-flotillin-HflC/K domain-containing protein 2; stomatin-prohibitin-flotillin-HflC/K domain-containing protein 2 {ECO:0000250; UniProtKB:Q8BFZ9, ECO:0000312; UniProtKB:Q8BFZ9}; UNQ2441; UNQ2441/PRO5003/PRO9924
Gene Symbols ERLIN2
Host Species Mouse
Immunogen E.coli-derived human Erlin-2/ERLIN2 recombinant protein (Position: D87-N339).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11160
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.