Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ERLIN2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597002
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597002 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597002 Supplier Invitrogen™ Supplier No. PA597002
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KIEEGHIGVY YRGGALLTST SGPGFHLMLP FITSYKSVQT TLQTDEVKNV PCGTSGGVMI YFDRIEVVNF LVPNAVYDIV KNYTADYDKA LIFNKIHHEL NQFCSVHTLQ EVYIELFGLE NDFSQESS; Positive Samples: U-937, U-251MG; Cellular Location: Endoplasmic reticulum membrane, Single-pass type II membrane protein.

ERLIN2, also named as C8orf2 and SPFH2, is a component of the ERLIN1/ERLIN2 complex which mediates the endoplasmic reticulum-associated degradation (ERAD) of inositol 1, 4, 5-trisphosphate receptors (IP3Rs) such as ITPR1. It promotes sterol-accelerated ERAD of HMGCR probably implicating an AMFR/gp78-containing ubiquitin ligase complex. ERLIN2 is involved in regulation of cellular cholesterol homeostasis by regulation the SREBP signaling pathway. The MW of ERLIN1/ERLIN2 complex is about 70 kDa.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ERLIN2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.57 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ERLIN2
Gene Accession No. O94905
Gene Alias BC036333; C87251; C8orf2; EMBL:AAI68668.1, ECO:0000312; EMBL:AAI68668.1}; endoplasmic reticulum lipid raft-associated protein 2; endoplasmic reticulum lipid raft-associated protein 2 {ECO:0000250; ER lipid raft associated 2; Erlin2; erlin-2; erlin-2 {ECO:0000250; erlin2 {ECO:0000312; HGNC:1356; MGC87072; NET32; RGD:1309010}; RGD1309010; SPFH domain family, member 2; SPFH domain-containing protein 2; SPFH domain-containing protein 2 {ECO:0000250; Spfh2; SPG18; stomatin-prohibitin-flotillin-HflC/K domain-containing protein 2; stomatin-prohibitin-flotillin-HflC/K domain-containing protein 2 {ECO:0000250; UniProtKB:Q8BFZ9, ECO:0000312; UniProtKB:Q8BFZ9}; UNQ2441; UNQ2441/PRO5003/PRO9924
Gene Symbols ERLIN2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 25-152 of human ERLIN2 (NP_0010037901).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11160
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.