Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ERLIN2 Recombinant Rabbit Monoclonal Antibody (JE57-03)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA544911
This item is not returnable.
View return policy
Description
ERLIN2 Recombinant Monoclonal Antibody for Western Blot, IHC (P)
ERLIN2, also named as C8orf2 and SPFH2, is a component of the ERLIN1/ERLIN2 complex which mediates the endoplasmic reticulum-associated degradation (ERAD) of inositol 1, 4, 5-trisphosphate receptors (IP3Rs) such as ITPR1. It promotes sterol-accelerated ERAD of HMGCR probably implicating an AMFR/gp78-containing ubiquitin ligase complex. ERLIN2 is involved in regulation of cellular cholesterol homeostasis by regulation the SREBP signaling pathway. The MW of ERLIN1/ERLIN2 complex is about 70 kDa.
Specifications
ERLIN2 | |
Recombinant Monoclonal | |
1 mg/mL | |
TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
O94905, Q8BFZ9 | |
ERLIN2 | |
Recombinant protein within human Erlin-2 aa 50-200 (Lumenal). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
JE57-03 | |
Unconjugated | |
ERLIN2 | |
BC036333; C87251; C8orf2; EMBL:AAI68668.1, ECO:0000312; EMBL:AAI68668.1}; endoplasmic reticulum lipid raft-associated protein 2; endoplasmic reticulum lipid raft-associated protein 2 {ECO:0000250; ER lipid raft associated 2; Erlin2; erlin-2; erlin-2 {ECO:0000250; erlin2 {ECO:0000312; HGNC:1356; MGC87072; NET32; RGD:1309010}; RGD1309010; SPFH domain family, member 2; SPFH domain-containing protein 2; SPFH domain-containing protein 2 {ECO:0000250; Spfh2; SPG18; stomatin-prohibitin-flotillin-HflC/K domain-containing protein 2; stomatin-prohibitin-flotillin-HflC/K domain-containing protein 2 {ECO:0000250; UniProtKB:Q8BFZ9, ECO:0000312; UniProtKB:Q8BFZ9}; UNQ2441; UNQ2441/PRO5003/PRO9924 | |
Rabbit | |
Protein A | |
RUO | |
11160, 244373 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction