Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ERMAP Antibody (OTIC8), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272339H
Description
ERMAP Monoclonal antibody specifically detects ERMAP in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence, CyTOFSpecifications
ERMAP | |
Monoclonal | |
HRP | |
PBS | |
Mouse | |
Immunogen affinity purified | |
RUO | |
114625 | |
Store at 4C in the dark. | |
IgG1 |
Flow Cytometry, Immunocytochemistry, CyTOF | |
OTIC8 | |
Flow Cytometry, Immunocytochemistry/ Immunofluorescence, CyTOF-ready | |
erythroblast membrane-associated protein, erythroblast membrane-associated protein (RD and SC blood groups), erythroblast membrane-associated protein (Scianna blood group), erythroid membrane-associated protein, hERMAP, MGC118810, MGC118811, PRO2801, Radin blood group, Radin blood group (Rd), Radin blood group antigen, RDMGC118812, Scianna blood group, Scianna blood group (Sc), Scianna blood group antigen, SCMGC118813 | |
Full length human recombinant protein of human ERMAP (NP_061008) produced in HEK293T cell. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction