Learn More
Invitrogen™ ERMAP Monoclonal Antibody (767505)

Description
Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.
Specifications
Specifications
| Antigen | ERMAP |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 767505 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene | ERMAP |
| Gene Accession No. | Q96PL5 |
| Gene Alias | AA409279; AI666418; BTN5; ERMAP; erythroblast membrane associated protein (Scianna blood group); erythroblast membrane-associated protein; erythroblast membrane-associated protein (Scianna blood group); erythroid membrane-associated protein; hERMAP; MGC118810; MGC118811; PRO2801; Radin blood group; Radin blood group (Rd); radin blood group antigen; RD; RDMGC118812; SC; Scianna blood group; Scianna blood group (Sc); scianna blood group antigen; SCMGC118813 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.