Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ERMAP Monoclonal Antibody (OTIC8)
GREENER_CHOICE

Catalog No. PIMA525042
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA525042 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA525042 Supplier Invitrogen™ Supplier No. MA525042
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ERMAP Monoclonal Antibody for ICC/IF, Flow

Erythroblast membrane-associated protein (ERMAP) is a gene situated on chromosome 1p34.2, encoding a protein primarily associated with erythroblast cells. ERMAP is expressed in erythroid lineage cells and plays a crucial role as an adhesion molecule, involved in the maturation and stability of the erythrocyte membrane. This protein is associated with the Scianna blood group system, and variations within the ERMAP gene lead to different antigenic expressions which are important in transfusion medicine. ERMAP is also significant in mediating cell-cell adhesion processes essential for erythropoiesis, where it facilitates receptor-ligand interactions in hematopoietic stem cells. Mutations or polymorphisms in ERMAP can result in altered blood group antigens, impacting blood transfusion compatibility and potentially leading to hemolytic reactions. Given its distinct role in erythrocyte formation, ERMAP is a subject of interest for further research into blood-related disorders and their genetic underpinnings.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ERMAP
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone OTIC8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ERMAP
Gene Accession No. Q96PL5
Gene Alias AA409279; AI666418; BTN5; ERMAP; erythroblast membrane associated protein (Scianna blood group); erythroblast membrane-associated protein; erythroblast membrane-associated protein (Scianna blood group); erythroid membrane-associated protein; hERMAP; MGC118810; MGC118811; PRO2801; Radin blood group; Radin blood group (Rd); radin blood group antigen; RD; RDMGC118812; SC; Scianna blood group; Scianna blood group (Sc); scianna blood group antigen; SCMGC118813
Gene Symbols ERMAP
Host Species Mouse
Immunogen Full length human recombinant protein of ERMAP produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114625
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.