Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ERMN Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544502
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544502 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544502 Supplier Invitrogen™ Supplier No. PA544502
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: RVIEFKKKHE EVSQFKEEGD ASEDSPLSSA SSQAVTPDEQ PTLGKKSDIS Sequence homology: Dog: 79%; Guinea Pig: 86%; Human: 100%; Rabbit: 86%; Rat: 86%.

ERMIN plays a role in cytoskeletal rearrangements during the late wrapping and/or compaction phases of myelinogenesis as well as in maintenance and stability of myelin sheath in the adult. ERMIN may play an important role in late-stage oligodendroglia maturation, myelin/Ranvier node formation during CNS development, and in the maintenance and plasticity of related structures in the mature CNS.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ERMN
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Ermn
Gene Accession No. Q8TAM6
Gene Alias A330104H05Rik; AI854460; ermin; ermin, ERM-like protein; Ermn; JN; juxtanodin; Kiaa1189; mKIAA1189; RGD1308367
Gene Symbols Ermn
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human KIAA1189.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57471
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.