Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ERMN Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558328
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558328 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558328 Supplier Invitrogen™ Supplier No. PA558328
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SLQETSADEM TFREGHQWEK IPLSGSNQEI RRQKERITEQ PLKEEEDEDR KNKGHQAAEI EWLGFRKPSQ AD Highest antigen sequence identity to the following orthologs: Mouse - 50%, Rat - 58%.

ERMIN plays a role in cytoskeletal rearrangements during the late wrapping and/or compaction phases of myelinogenesis as well as in maintenance and stability of myelin sheath in the adult. ERMIN may play an important role in late-stage oligodendroglia maturation, myelin/Ranvier node formation during CNS development, and in the maintenance and plasticity of related structures in the mature CNS.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ERMN
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Ermn
Gene Accession No. Q8TAM6
Gene Alias A330104H05Rik; AI854460; ermin; ermin, ERM-like protein; Ermn; JN; juxtanodin; Kiaa1189; mKIAA1189; RGD1308367
Gene Symbols Ermn
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ERMN. Recombinant protein control fragment (Product #RP-96088).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57471
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.