Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ERMP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA551191
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA551191 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA551191 Supplier Invitrogen™ Supplier No. PA551191
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total ERMP1 protein.

Within the ovary, required for the organization of somatic cells and oocytes into discrete follicular structures.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ERMP1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene ERMP1
Gene Accession No. Q7Z2K6
Gene Alias aminopeptidase Fxna; bA207C16.3; bA207C16.3 (novel protein similar to predicted yeast, plant and worm proteins); D19Ertd410e; D19Wsu12e; Endoplasmic reticulum metallopeptidase 1; Ermp1; Felix-ina; Fxna; KIAA1815; mKIAA1815; putative aminopeptidase Fxna
Gene Symbols ERMP1
Host Species Rabbit
Immunogen Fusion protein of human ERMP1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79956
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.