Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ERN1/ERN2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115434
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115434 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115434 Supplier Invitrogen™ Supplier No. PA5115434
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total IRE1.

ERN1 (Endoplasmic Reticulum To Nucleus Signaling 1) is a Protein Coding gene. Diseases associated with ERN1 include Arrhythmogenic Right Ventricular Dysplasia, Familial, 5 and Epiphyseal Dysplasia, Multiple, With Early-Onset Diabetes Mellitus. Among its related pathways are Spinocerebellar ataxia and Photodynamic therapy-induced unfolded protein response. Gene Ontology (GO) annotations related to this gene include protein homodimerization activity and transferase activity, transferring phosphorus-containing groups. An important paralog of this gene is ERN2. This gene encodes the transmembrane protein kinase inositol-requiring enzyme 1. The encoded protein contains two functional catalytic domains, a serine/threonine-protein kinase domain and an endoribonuclease domain. This protein functions as a sensor of unfolded proteins in the endoplasmic reticulum (ER) and triggers an intracellular signaling pathway termed the unfolded protein response (UPR). The UPR is an ER stress response that is conserved from yeast to mammals and activates genes involved in degrading misfolded proteins, regulating protein synthesis and activating molecular chaperones. This protein specifically mediates the splicing and activation of the stress response transcription factor X-box binding protein 1. [provided by RefSeq, Aug 2017].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ERN1/ERN2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene Ern1
Gene Accession No. O75460, Q76MJ5, Q9EQY0, Q9Z2E3
Gene Alias 9030414B18Rik; AI225830; C85377; endoplasmic reticulum (ER) to nucleus signalling 1; endoplasmic reticulum (ER) to nucleus signalling 2; endoplasmic reticulum to nucleus signaling 1; endoplasmic reticulum to nucleus signaling 2; endoplasmic reticulum to nucleus signalling 2; endoplasmic reticulum-to-nucleus signaling 1; Endoplasmic reticulum-to-nucleus signaling 2; Endoribonuclease; ER to nucleus signalling 1; ER to nucleus signalling 2; ERN1; ERN2; hIRE1p; hIRE2p; inositol-requiring 1; inositol-requiring 1 (Yeast homologue); inositol-requiring 1 alpha; inositol-requiring 1 beta; inositol-requiring enzyme 1; inositol-requiring protein 1; Inositol-requiring protein 2; Ire1; IRE1 beta; IRE1, S. cerevisiae, homolog of; Ire1a; Ire1alpha; ire1-alpha; IRE1b; Ire1-beta; Ire1p; Ire2; IRE2p; mIre1; protein kinase/endoribonuclease; RGD1559716; Serine/threonine-protein kinase; Serine/threonine-protein kinase/endoribonuclease IRE1; serine/threonine-protein kinase/endoribonuclease IRE2
Gene Symbols Ern1, Ern2
Host Species Rabbit
Immunogen A synthesized peptide derived from human ERN1(Accession O75460), corresponding to amino acid residues P603-V653.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10595, 2081, 26918, 365363, 498013, 78943
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.