Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ERN2 Polyclonal Antibody
ERN2 Polyclonal Antibody
Supplier: Thermo Scientific PA5116228
Description
ERN2 Polyclonal Antibody for Western Blot

Specifications
ERN2 | |
Polyclonal | |
Unconjugated | |
ERN2 | |
endoplasmic reticulum (ER) to nucleus signalling 2; endoplasmic reticulum to nucleus signaling 2; endoplasmic reticulum to nucleus signalling 2; endoplasmic reticulum-to-nucleus signaling 2; Endoribonuclease; ER to nucleus signalling 2; Ern1; ern2; hIRE2p; inositol-requiring 1 (Yeast homologue); inositol-requiring 1 beta; inositol-requiring protein 2; Ire1; IRE1 beta; IRE1, S. cerevisiae, homolog of; Ire1b; ire1-beta; IRE2; IRE2p; mIre1; Serine/threonine-protein kinase; Serine/threonine-protein kinase/endoribonuclease IRE2 | |
Rabbit | |
Antigen Affinity Chromatography, Protein A | |
RUO | |
26918 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q9Z2E3 | |
ERN2 | |
KLH conjugated synthetic peptide between 883-911 amino acids from the C-terminal region of mouse Ern2. | |
400 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction