Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ERO1L Antibody (V23P3C9*H2), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250388B
Description
ERO1L Monoclonal specifically detects ERO1L in Human, Mouse, Rat, Porcine, Canine, Equine, Sheep samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Specifications
ERO1L | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
ERO1A | |
Peptide Sequence - KLIANMPESG (peptide immunogen is identical in rat (Rattus norvegicus), mouse (Mus musculus), sheep (Ovis aries), bovine (Bos taurus), canine (Canis familiaris), porcine (Sus scrofa) and equine (Equus caballus) | |
0.1 mL | |
Cancer, ER Markers, Hypoxia, Membrane Trafficking and Chaperones, Signal Transduction | |
30001 | |
Store at 4C in the dark. | |
IgG1 κ |
Western Blot, ELISA, Immunohistochemistry | |
V23P3C9*H2 | |
Western Blot, ELISA, Immunohistochemistry | |
EC 1.8.4, EC 1.8.4.-, Endoplasmic oxidoreductin-1-like protein, ERO1 (S. cerevisiae)-like, ERO1A, ERO1-alpha, ERO1-L, ERO1-L-alpha, ERO1-like (S. cerevisiae), ERO1-like protein alpha, oxidoreductin-1-L-alpha | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Pig, Canine, Equine, Sheep | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction