Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    ERO1L Antibody (V23P3C9*H2), mFluor Violet 610 SE, Novus Biologicals™
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250388MFV610
Description
ERO1L Monoclonal antibody specifically detects ERO1L in Human, Mouse, Rat, Porcine, Canine, Equine, Sheep samples. It is validated for Western Blot, ELISA, ImmunohistochemistrySpecifications
| ERO1L | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Pig, Canine | |
| Purified | 
| Western Blot, ELISA, Immunohistochemistry | |
| V23P3C9*H2 | |
| Western Blot, ELISA, Immunohistochemistry | |
| EC 1.8.4, EC 1.8.4.-, Endoplasmic oxidoreductin-1-like protein, ERO1 (S. cerevisiae)-like, ERO1A, ERO1-alpha, ERO1-L, ERO1-L-alpha, ERO1-like (S. cerevisiae), ERO1-like protein alpha, oxidoreductin-1-L-alpha | |
| Peptide Sequence - KLIANMPESG (peptide immunogen is identical in rat (Rattus norvegicus), mouse (Mus musculus), sheep (Ovis aries), bovine (Bos taurus), canine (Canis familiaris), porcine (Sus scrofa) and equine (Equus caballus) | |
| 0.1 mL | |
| Cancer, ER Markers, Hypoxia, Membrane Trafficking and Chaperones, Signal Transduction | |
| 30001 | |
| Store at 4C in the dark. | |
| IgG1 κ | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction