Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ERO1LB Antibody (M37-P5D11), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250389AF405
Description
ERO1LB Monoclonal specifically detects ERO1LB in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
ERO1LB | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
ERO1B | |
Peptide - LAPSRGEDDG | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
M37-P5D11 | |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
DKFZp779C1042, DKFZp779I0141, EC 1.8.4, EC 1.8.4.-, Endoplasmic oxidoreductin-1-like protein B, ERO1-L(beta), ERO1-L-beta, ERO1-like beta (S. cerevisiae), ERO1-like protein beta, FLJ11003, oxidoreductin-1-L-beta | |
Mouse | |
Protein A purified | |
RUO | |
56605 | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction