Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ERO1LB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113082
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113082 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113082 Supplier Invitrogen™ Supplier No. PA5113082
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ERO1LB Polyclonal Antibody for Western Blot, IHC (P), ELISA

ERO1LB is essential oxidoreductase that oxidizes proteins in the endoplasmic reticulum to produce disulfide bonds. ERO1LB acts by oxidizing directly P4HB/PDI isomerase through a direct disulfide exchange. ERO1LB does not act as a direct oxidant of folding substrate, but relies on P4HB/PDI to transfer oxidizing equivalent.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ERO1LB
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene Ero1b
Gene Accession No. Q86YB8, Q8R2E9
Gene Alias 1300013B24Rik; 1700065B09Rik; AI447560; endoplasmic oxidoreductase 1 beta; endoplasmic oxidoreductin-1-like protein B; endoplasmic reticulum oxidoreductase 1 beta; endoplasmic reticulum oxidoreductase beta; Endoplasmic reticulum oxidoreductin-1-like protein B; Ero1b; Ero1beta; ero1-beta; ERO1LB; ERO1-L-beta; ERO1-like beta; ERO1-like beta (S. cerevisiae); ERO1-like protein beta; Oxidoreductin-1-L-beta; RGD1563548
Gene Symbols Ero1b, Ero1lb
Host Species Rabbit
Immunogen Recombinant Human ERO1-like protein beta protein (101-400AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56605, 67475
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.