Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ERO1LB Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS14627R

Description
ERO1LB is essential oxidoreductase that oxidizes proteins in the endoplasmic reticulum to produce disulfide bonds. ERO1LB acts by oxidizing directly P4HB/PDI isomerase through a direct disulfide exchange. ERO1LB does not act as a direct oxidant of folding substrate, but relies on P4HB/PDI to transfer oxidizing equivalent.Specifications
ERO1LB | |
Polyclonal | |
Unconjugated | |
Ero1b | |
1300013B24Rik; 1700065B09Rik; AI447560; endoplasmic oxidoreductase 1 beta; endoplasmic oxidoreductin-1-like protein B; endoplasmic reticulum oxidoreductase 1 beta; Endoplasmic reticulum oxidoreductase beta; Endoplasmic reticulum oxidoreductin-1-like protein B; ERO1B; Ero1beta; ero1-beta; Ero1lb; ERO1-L-beta; ERO1-like beta; ERO1-like beta (S. cerevisiae); ERO1-like protein beta; Oxidoreductin-1-L-beta; RGD1563548 | |
Rabbit | |
Protein A | |
RUO | |
56605 | |
-20°C | |
Liquid |
Western Blot | |
1 μg/mL | |
TBS with 1% BSA, 50% glycerol and 0.03% Proclin 300; pH | |
Q86YB8 | |
Ero1b | |
KLH conjugated synthetic peptide derived from human ERO1LB. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction