Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ERP29 Antibody, DyLight 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299200V
Description
ERP29 Polyclonal specifically detects ERP29 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
ERP29 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
C12orf8chromosome 12 open reading frame 8, endoplasmic reticulum lumenal protein ERp28, endoplasmic reticulum protein 29, Endoplasmic reticulum resident protein 28, endoplasmic reticulum resident protein 29, ERp28Endoplasmic reticulum resident protein 31, ERp29, ERp31ERP28, PDIA9, PDI-DB, protein disulfide isomerase family A, member 9 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human ERP29. | |
0.1 mL | |
Core ESC Like Genes, Membrane Trafficking and Chaperones, Stem Cell Markers | |
10961 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 405 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction