Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ERP29 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP133076
Description
ERP29 Polyclonal specifically detects ERP29 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
ERP29 | |
Polyclonal | |
Unconjugated | |
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
ERP29 | |
Recombinant protein encompassing a sequence within the center region of human ERP29. The exact sequence is proprietary. | |
100 μL | |
Core ESC Like Genes, Membrane Trafficking and Chaperones, Stem Cell Markers | |
10961 | |
Human, Mouse, Rat | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
1.0mg/mL | |
Western Blot 1:500-1:3000, Immunohistochemistry 10 - 1:500, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
C12orf8chromosome 12 open reading frame 8, endoplasmic reticulum lumenal protein ERp28, endoplasmic reticulum protein 29, Endoplasmic reticulum resident protein 28, endoplasmic reticulum resident protein 29, ERp28Endoplasmic reticulum resident protein 31, ERp29, ERp31ERP28, PDIA9, PDI-DB, protein disulfide isomerase family A, member 9 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Tested in human. Not yet tested in any other species. | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction