Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ERP29 Recombinant Rabbit Monoclonal Antibody (1J0D3)
GREENER_CHOICE

Catalog No. PIMA537989
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537989 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA537989 Supplier Invitrogen™ Supplier No. MA537989
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Samples: 293T, HepG2, A-431, NIH/3T3, Mouse brain, Mouse kidney, Rat lung, Rat spleen, Rat kidney Immunogen sequence: MAAAVPRAAF LSPLLPLLLG FLLLSAPHGG SGLHTKGALP LDTVTFYKVI PKSKFVLVKF DTQYPYGEKQ DEFKRLAENS ASSDDLLVAE VGISDYGDKL NM.

This gene encodes a reticuloplasmin, a protein which resides in the lumen of the endoplasmic reticulum. The protein shows sequence similarity to the protein disulfide isomerase family. However, it lacks the thioredoxin motif characteristic of this family, suggesting that this protein does not function as a disulfide isomerase. The protein dimerizes and is thought to play a role in the processing of secretory proteins within the ER. Alternative splicing results in multiple transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ERP29
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone 1J0D3
Concentration 1.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Erp29
Gene Accession No. P30040, P52555, P57759
Gene Alias 1200015M03Rik; 2810446M09Rik; AW209030; C12orf8; endoplasmic reticulum lumenal protein ERp28; endoplasmic reticulum protein 29; endoplasmic reticulum protein ERp29; endoplasmic reticulum resident protein 28; Endoplasmic reticulum resident protein 29; endoplasmic reticulum resident protein 31; endoplasmic retuclum protein 29; epididymis secretory protein Li 107; Erp28; ERp29; Erp31; HEL-S-107; PDIA9; PDI-DB; protein disulfide isomerase family A, member 9
Gene Symbols Erp29
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-102 of human ERp29 (P30040).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10961, 117030, 67397
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.