Learn More
Invitrogen™ ERP29 Recombinant Rabbit Monoclonal Antibody (JG34-42)

Description
Positive Control: THP-1, F9, PMVEC, SiHa, rat testis tissue, human thyroid gland tissue, human colon cancer tissue.
Specifications
Specifications
| Antigen | ERP29 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | JG34-42 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | Erp29 |
| Gene Accession No. | P30040 |
| Gene Alias | 1200015M03Rik; 2810446M09Rik; AW209030; C12orf8; endoplasmic reticulum lumenal protein ERp28; endoplasmic reticulum protein 29; endoplasmic reticulum protein ERp29; endoplasmic reticulum resident protein 28; Endoplasmic reticulum resident protein 29; endoplasmic reticulum resident protein 31; endoplasmic retuclum protein 29; epididymis secretory protein Li 107; Erp28; ERp29; Erp31; HEL-S-107; PDIA9; PDI-DB; protein disulfide isomerase family A, member 9 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.