Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ERP44 Antibody, Janelia Fluor™ 549, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297000JF549
Description
ERP44 Polyclonal antibody specifically detects ERP44 in Human samples. It is validated for Western Blot, ELISA, ImmunoprecipitationSpecifications
ERP44 | |
Polyclonal | |
Western Blot, ELISA, Immunoprecipitation | |
endoplasmic reticulum protein 44, endoplasmic reticulum resident protein 44 kDa, ER protein 44, ERp44, KIAA0573endoplasmic reticulum resident protein 44, PDIA10, protein disulfide isomerase family A, member 10, thioredoxin domain containing 4 (endoplasmic reticulum), Thioredoxin domain-containing protein 4, TXNDC4 | |
Produced in rabbits immunized with purified, recombinant Human ERP44 (Accession#: NP_055866.1; Met1-Asp402) | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, ELISA, Immunoprecipitation | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
23071 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction