Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ERP44 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533519
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533519 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533519 Supplier Invitrogen™ Supplier No. PA533519
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ERP44 Polyclonal Antibody for IHC (P)

Mediates thiol-dependent retention in the early secretory pathway, forming mixed disulfides with substrate proteins through its conserved CRFS motif. Inhibits the calcium channel activity of ITPR1. May have a role in the control of oxidative protein folding in the endoplasmic reticulum. Required to retain ERO1L and ERO1LB in the endoplasmic reticulum.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ERP44
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene ERP44
Gene Accession No. Q3T0L2, Q9BS26, Q9D1Q6
Gene Alias 1110001E24Rik; AI849526; AL033348; BWK4; endoplasmic reticulum protein 44; endoplasmic reticulum resident protein 44; endoplasmic reticulum resident protein 44 kDa; endoplasmic reticulum resident protein 44kDa; endoplasmic reticulum resident protein ERp44; ER protein 44; Erp44; KIAA0573; OTTHUMP00000063799; PDIA10; protein disulfide isomerase family A, member 10; thioredoxin domain containing 4 (endoplasmic reticulum); thioredoxin domain-containing protein 4; Txndc4; UNQ532/PRO1075
Gene Symbols ERP44
Host Species Rabbit
Immunogen Synthetic 20 amino acid peptide from internal region of human ERP44 / TXNDC4.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100054749, 100192437, 100344280, 100716079, 23071, 298066, 420994, 474781, 506157, 76299
Target Species Human, Mouse, Rat, Canine, Rabbit, Monkey, Bovine, Pig, Horse, Chicken, Guinea Pig, Frog, Bat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.