Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ERP44 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5112156
Description
This product can be used in direct ELISA.
Mediates thiol-dependent retention in the early secretory pathway, forming mixed disulfides with substrate proteins through its conserved CRFS motif. Inhibits the calcium channel activity of ITPR1. May have a role in the control of oxidative protein folding in the endoplasmic reticulum. Required to retain ERO1L and ERO1LB in the endoplasmic reticulum.
Specifications
ERP44 | |
Polyclonal | |
Unconjugated | |
ERP44 | |
1110001E24Rik; AI849526; AL033348; BWK4; endoplasmic reticulum protein 44; endoplasmic reticulum resident protein 44; endoplasmic reticulum resident protein 44 kDa; endoplasmic reticulum resident protein 44kDa; endoplasmic reticulum resident protein ERp44; ER protein 44; Erp44; KIAA0573; OTTHUMP00000063799; PDIA10; protein disulfide isomerase family A, member 10; thioredoxin domain containing 4 (endoplasmic reticulum); thioredoxin domain-containing protein 4; Txndc4; UNQ532/PRO1075 | |
Rabbit | |
Antigen Affinity Chromatography, Protein A | |
RUO | |
23071 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunoprecipitation, Western Blot | |
1 mg/mL | |
PBS with no preservative | |
Q9BS26 | |
ERP44 | |
Recombinant Human ERp44/TXNDC4 Protein. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction