Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ERp57 Monoclonal Antibody (OTI4D7)
GREENER_CHOICE

Catalog No. PIMA526027
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526027 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526027 Supplier Invitrogen™ Supplier No. MA526027
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ERp57 Monoclonal Antibody for Western Blot, IHC (P)

This gene encodes a protein of the endoplasmic reticulum that interacts with lectin chaperones calreticulin and calnexin to modulate folding of newly synthesized glycoproteins. The protein was once thought to be a phospholipase; however, it has been demonstrated that the protein actually has protein disulfide isomerase activity. It is thought that complexes of lectins and this protein mediate protein folding by promoting formation of disulfide bonds in their glycoprotein substrates.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ERp57
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI4D7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PDIA3
Gene Accession No. P30101
Gene Alias 58 kDa glucose-regulated protein; 58 kDa microsomal protein; 58kDa; Disulfide isomerase ER-60; endoplasmic reticulum P58; endoplasmic reticulum protein; endoplasmic reticulum resident protein 57; Endoplasmic reticulum resident protein 60; epididymis secretory protein Li 269; epididymis secretory sperm binding protein Li 93n; ER p57; ER protein 57; ER protein 60; ER60; ER-60 protease; Erp; ERp 57; ERp57; ERP57 protein; ERP60; ERp61; glucose regulated protein 58; glucose regulated protein 58kD; glucose regulated protein, 58 kDa; glucose regulated protein, 58kDa; GRP 57; GRP 58; GRP57; Grp58; grp-58; HEL-S-269; HEL-S-93n; HIP-70; HsT17083; I79_003765; oxidoreductase ERp57; p58; PDI; PDIA 3; PDIA3; PDI-Q2; phospholipase C, alpha; phospholipase C-alpha; PI PLC; PI-PLC; PLC alpha; PLC[a]; Plca; protein disulfide isomerase associated 3; protein disulfide isomerase family A member 3; protein disulfide isomerase family A, member 3; protein disulfide isomerase-associated 3; protein disulfide-isomerase A3; Q-2
Gene Symbols PDIA3
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 140-505 of PDIA3 produced in E.coli.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2923
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.