Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ERp57/PDIA3 Antibody (4F9), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259695UV
Description
ERp57/PDIA3 Monoclonal specifically detects ERp57/PDIA3 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
ERp57/PDIA3 | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
PDIA3 | |
Human recombinant ERp57 (Grp58) protein corresponding to amino acids 25 - 505 (his-tagged) | |
100 μL | |
Cellular Markers, ER Markers, Membrane Trafficking and Chaperones | |
2923 | |
Human | |
Purified |
Western Blot, Immunocytochemistry, Immunofluorescence | |
4F9 | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
Disulfide isomerase ER-60, EC 5.3.4.1, endoplasmic reticulum P58, Endoplasmic reticulum resident protein 57, Endoplasmic reticulum resident protein 60, ER protein 60, ER60, ERP57, ERp5758 kDa glucose-regulated protein, ERP60, ERp6058 kDa microsomal protein, ERp61, glucose regulated protein, 58kDa, GRP57, GRP58ER protein 57, HsT17083, P58, phospholipase C-alpha, PI-PLC, protein disulfide isomerase family A, member 3, protein disulfide isomerase-associated 3, protein disulfide-isomerase A3 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Detects approx 110kDa. | |
Store at 4C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction