Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ERp72 Antibody (OTI2B9), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271488FR
Description
ERp72 Monoclonal specifically detects ERp72 in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Western Blot, Immunohistochemistry.Specifications
ERp72 | |
Monoclonal | |
DyLight 680 | |
PBS with 0.05% Sodium Azide | |
PDIA4 | |
Full length recombinant protein of human ERp72 (NP_004902) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Canine, Primate, Monkey | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI2B9 | |
Western Blot, Immunohistochemistry | |
Endoplasmic reticulum resident protein 70, Endoplasmic reticulum resident protein 72, ER protein 70, ERp70, ERP70ER protein 72, ERp72, ERp-72, ERP72EC 5.3.4.1, intestinal-related), protein disulfide isomerase family A, member 4, protein disulfide isomerase related protein (calcium-binding protein, protein disulfide isomerase-associated 4, protein disulfide-isomerase A4 | |
Mouse | |
Affinity Purified | |
Core ESC Like Genes, Membrane Trafficking and Chaperones, Stem Cell Markers | |
9601.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction