Learn More
Invitrogen™ ERP72 Monoclonal Antibody (OTI2E2)

Description
ERP72 Monoclonal Antibody for Western Blot, ICC/IF, Flow
Specifications
Specifications
| Antigen | ERP72 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | OTI2E2 |
| Concentration | 0.87 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | PDIA4 |
| Gene Accession No. | P13667 |
| Gene Alias | AI987846; caBP2; Cai; calcium binding protein, intestinal; calcium-binding protein 2; EC 5.3.4.1; endoplasmic reticulum protein 72; endoplasmic reticulum resident protein 70; Endoplasmic reticulum resident protein 72; ER protein 70; ER protein 72; Erp70; Erp72; ERp-72; PDIA4; protein disu; protein disulfide isomerase A4; protein disulfide isomerase associated 4; protein disulfide isomerase family A member 4; protein disulfide isomerase family A, member 4; protein disulfide isomerase related protein (calcium-binding protein, intestinal-related); protein disulfide isomerase-associated 4; protein disulfide-isomerase A4; Protein ERp-72; testicular tissue protein Li 137 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.