Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ERp72 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5122011
This item is not returnable.
View return policy
Description
ERp72 Polyclonal Antibody for Western Blot, IHC (P)
ERp72 is an endoplasmic reticulum luminal protein that is both a stress protein and a member of the protein disulfide isomerase family of proteins. Sequence analysis of ERp72 shows that it shares sequence identity with PDI (protein disulphide isomerase) at three discrete regions, as well as containing the ER retention signal KEEL at its C-terminus. ERp72 is shown to have three copies of sequence believed to be CGHC-containing active sites, which are also found in PDI. These CGHC sites are critical sites that are involved in the protein disulphide isomerization process. These results suggest that ERp72 is a newly described member of the family of CGHC-containing proteins that functions as a molecular chaperone.
Specifications
ERp72 | |
Polyclonal | |
Unconjugated | |
PDIA4 | |
AI987846; caBP2; Cai; calcium binding protein, intestinal; calcium-binding protein 2; EC 5.3.4.1; endoplasmic reticulum protein 72; endoplasmic reticulum resident protein 70; Endoplasmic reticulum resident protein 72; ER protein 70; ER protein 72; Erp70; Erp72; ERp-72; PDIA4; protein disu; protein disulfide isomerase A4; protein disulfide isomerase associated 4; protein disulfide isomerase family A member 4; protein disulfide isomerase family A, member 4; protein disulfide isomerase related protein (calcium-binding protein, intestinal-related); protein disulfide isomerase-associated 4; protein disulfide-isomerase A4; Protein ERp-72; testicular tissue protein Li 137 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9601 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
500 μg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
P13667 | |
PDIA4 | |
Recombinant protein Disulfide Isomerase A4. The antigen corresponds to amino acid range 505-636 of the target protein. | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction