Learn More
Invitrogen™ ERp72 Polyclonal Antibody, Alexa Fluor™ 488

Description
PA1007A488 detects ERp72 protein in human and mouse samples. The PA1007A488 immunogen is a synthetic peptide corresponding to residues K(629) F I E E H A T K L S R T K E E L(645) of human ERp72. This peptide (Cat. # PEP-223) is available for use in neutralization and control experiments.
Specifications
Specifications
| Antigen | ERp72 |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Alexa Fluor 488 |
| Formulation | PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.4 |
| Gene | PDIA4 |
| Gene Accession No. | P08003, P13667 |
| Gene Alias | AI987846; caBP2; Cai; calcium binding protein, intestinal; calcium-binding protein 2; EC 5.3.4.1; endoplasmic reticulum protein 72; endoplasmic reticulum resident protein 70; Endoplasmic reticulum resident protein 72; ER protein 70; ER protein 72; Erp70; Erp72; ERp-72; PDIA4; protein disu; protein disulfide isomerase A4; protein disulfide isomerase associated 4; protein disulfide isomerase family A member 4; protein disulfide isomerase family A, member 4; protein disulfide isomerase related protein (calcium-binding protein, intestinal-related); protein disulfide isomerase-associated 4; protein disulfide-isomerase A4; Protein ERp-72; testicular tissue protein Li 137 |
| Gene Symbols | PDIA4 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.