Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ERR alpha Mouse anti-Human, Unconjugated, Clone: 2ERR10, MilliporeSigma™

Catalog No. MABE18911MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE18911MI 100 μg
MABE18912MI 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABE18911MI Supplier MilliporeSigma Supplier No. MABE1891100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ERR alpha Monoclonal antibody specifically detects ERR alpha in Human samples. It is validated for Chromatin Immunoprecipitation (ChIP), ELISA, Immunocytochemistry, Immunoprecipitation, Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ERR alpha
Applications ChIP Assay, ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 2ERR10
Conjugate Unconjugated
Gene Accession No. NP_001269379
Gene Alias Steroid hormone receptor ERR1;Estrogen receptor-like 1;Estrogen-related receptor alpha;Nuclear receptor subfamily 3 group B member 1
Gene Symbols ESRRA, ERR1, ESRL1, NR3B1
Host Species Mouse
Immunogen His-tagged full length human recombinant Steroid hormone receptor ERR alpha (ERR1).
Molecular Weight of Antigen ∼53 kDa observed; 45.51 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Protein G Purified
Quantity 100 μg
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Test Specificity Clone 2ERR10 is a mouse monoclonal antibody that detects human Steroid hormone receptor ERR alpha (ERR1).
Target Species Human
Content And Storage Stable for 1 year at 2 to 8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.