Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ERR gamma/NR3B3 Antibody (OTI1E5), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270691PCP
Description
ERR gamma/NR3B3 Monoclonal antibody specifically detects ERR gamma/NR3B3 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
ERR gamma/NR3B3 | |
Monoclonal | |
PerCP | |
PBS | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry | |
OTI1E5 | |
Western Blot, Immunohistochemistry | |
DKFZp781L1617, ERR gamma-2, ERR3, ERRG2, ERRgamma, Estrogen receptor-related protein 3, estrogen-related receptor gamma, FLJ16023, KIAA0832, NR3B3, Nuclear receptor subfamily 3 group B member 3 | |
Human recombinant protein fragment corresponding to amino acids 24-263 of human ESRRG(NP_001429) produced in E.coli. | |
0.1 mL | |
Cancer, Endocrinology, Signal Transduction | |
2104 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction