Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ERR gamma/NR3B3 Antibody (SR1899), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32230350UL
Description
ERR gamma/NR3B3 Monoclonal antibody specifically detects ERR gamma/NR3B3 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
ERR gamma/NR3B3 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot | |
SR1899 | |
Western Blot 1:500-1:2000 | |
DKFZp781L1617, ERR gamma-2, ERR3, ERRG2, ERRgamma, Estrogen receptor-related protein 3, estrogen-related receptor gamma, FLJ16023, KIAA0832, NR3B3, Nuclear receptor subfamily 3 group B member 3 | |
A synthesized peptide derived from human ERR gamma/NR3B3 (Uniprot #: P62508) | |
50 μg | |
Cancer, Endocrinology, Signal Transduction | |
2104 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction