Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ERR gamma/NR3B3 Rabbit anti-Human, Mouse, Rat, Clone: 2W3A9, Novus Biologicals™
SDP

Catalog No. p-7248324 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB168761 20 μg
NB168760 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB168761 Supplier Novus Biologicals Supplier No. NBP31683620UL
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

ERR gamma/NR3B3 Monoclonal antibody specifically detects ERR gamma/NR3B3 in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen ERR gamma/NR3B3
Applications Western Blot
Classification Monoclonal
Clone 2W3A9
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias DKFZp781L1617, ERR gamma-2, ERR3, ERRG2, ERRgamma, Estrogen receptor-related protein 3, estrogen-related receptor gamma, FLJ16023, KIAA0832, NR3B3, Nuclear receptor subfamily 3 group B member 3
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human ERR gamma/NR3B3 (P62508).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Cancer, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 2104
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.