Learn More
Invitrogen™ ERRF Polyclonal Antibody

Description
Immunogen sequence: HLTRVTPMGG GCLAQARATL PLCRGSVASA SFPVSPLCPQ EVPEAKGKPV KAAPVRSSTW GTVKDSLKAL SSCVCGQ Highest antigen sequence identity to the following orthologs: Mouse - 42%, Rat - 30%.
Specifications
Specifications
| Antigen | ERRF |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | SRARP |
| Gene Accession No. | Q8NEQ6 |
| Gene Alias | C1orf64; chromosome 1 open reading frame 64; ER-related factor; ERRF; Gm694; predicted gene 694; RP11-5P18.4; Srarp; SSPR; Steroid receptor-associated and regulated protein; Steroid receptor-regulated protein; uncharacterized protein C1orf64; uncharacterized protein C1orf64 homolog |
| Gene Symbols | SRARP |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.