Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Erucamide 85.0+%, TCI America™
SDP

Catalog No. D1008100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1008100G 100 g
D100825G 25 g
D1008500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D1008100G Supplier TCI America Supplier No. D1008100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 112-84-5
Molecular Formula C22H43NO
Molecular Weight (g/mol) 337.59
MDL Number MFCD00882379
InChI Key UAUDZVJPLUQNMU-KTKRTIGZSA-N
Synonym erucamide, erucylamide, z-docos-13-enamide, erucyl amide, cis-13-docosenoamide, erucic acid amide, 13-docosenamide, z, cis-13-docosenamide, 13-docosenamide, unii-0v89vy25bn
PubChem CID 5365371
IUPAC Name (13Z)-docos-13-enamide
SMILES CCCCCCCC\C=C/CCCCCCCCCCCC(N)=O

Specifications

Melting Point 81°C
Color White
Quantity 100 g
Formula Weight 337.59
Percent Purity ≥85.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Erucamide
EINECSNumber (2)-0824&(2)-0981
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.