Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Erucic Acid 85.0+%, TCI America™
SDP

Catalog No. D0965400G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
400 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0965400G 400 g
D096525G 25 g
D0965100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D0965400G Supplier TCI America Supplier No. D0965400G

Chemical Identifiers

CAS 112-86-7
Molecular Formula C22H42O2
Molecular Weight (g/mol) 338.576
MDL Number MFCD00063188
InChI Key DPUOLQHDNGRHBS-KTKRTIGZSA-N
Synonym erucic acid, cis-13-docosenoic acid, z-docos-13-enoic acid, 13-docosenoic acid, z, cis-erucic acid, 13-cis-docosenoic acid, 13-docosenoic acid, z-13-docosenoic acid, hystrene 2290
PubChem CID 5281116
ChEBI CHEBI:28792
IUPAC Name (Z)-docos-13-enoic acid
SMILES CCCCCCCCC=CCCCCCCCCCCCC(=O)O

Specifications

Color White-Yellow
Quantity 400 g
Formula Weight 338.58
Percent Purity ≥85.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Erucic Acid
EINECSNumber (2)-0609&(2)-0983
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.