Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Erucic acid, ≥99.0% (GC), MilliporeSigma™ Supelco™

Catalog No. p-7187671
Click to view available options
Quantity:
1 g
5 g

Specifications

CAS 29232-93-7
Flash Point 113°C
Physical Form Neat
Linear Formula CH3(CH2)7 CH=CH(CH2)11 COOH
MDL Number MFCD00063188
Melting Point 28°C to 32°C
Boiling Point 358°C (400 mmHg)
Synonym cis-13-Docosenoic acid; Prifac 2990
Type Analytical Standard
Quantity 1 g
Formula Weight 338.57
Percent Purity ≥99.0% (GC)
Grade Analytical Standard
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.