Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ERV3-1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00
Specifications
Antigen | ERV3-1 |
---|---|
Dilution | Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
ERV3-1 Polyclonal antibody specifically detects ERV3-1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
ERV3-1 | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human | |
Endogenous Retroviral Sequence 3, Endogenous Retroviral Sequence 3 (Includes Zinc Finger Protein H-Plk/HPF9), Endogenous Retrovirus Group 3 Member 1, Endogenous Retrovirus Group 3 Member 1 Env Polyprotein, Endogenous Retrovirus Group 3 Member 1, Envelope, Endogenous Retrovirus Group 3, Member 1, envelope polyprotein, ERV3, ERV3 Envelope Protein, ERV-3 Envelope Protein, ERV3-1 Envelope Protein, ERV-R, ERV-R Envelope Protein, HERVR, HERV-R, HERV-R_7q21.2 Provirus Ancestral Env Polyprotein | |
This antibody was developed against a recombinant protein corresponding to amino acids: VCYDPKSSPGIWFEIHVGSKEGDLLNQTKVFPSGKDVVSLYFDVCQIVSMGSLFPVIFSSMEYYSSCHKNRYAHPACSTDSPVTTCWDCTTWSTNQQSLGPIMLTKIPLEPDCKTSTCNSVNLTILEPDQPIWTTGLKAPL | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
Polyclonal | |
Purified | |
RUO | |
PBS (pH 7.2) and 40% Glycerol | |
2086 | |
IgG | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title