Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ERV3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582579
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582579 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582579 Supplier Invitrogen™ Supplier No. PA582579
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QLAENIASSL HVASCYVCGG MNMGDQWPWE ARELMPQDNF TLTASSLEPA PSSQSIWFLK TSIIGKFCIA RWGKAFTDPV GELTCLGQQY YNETLGKTLW RGKSNNSESP HPSPFSRFPS LNHSWYQLEA Highest antigen sequence indentity to the following orthologs: Mouse - 19%, Rat - 24%.

Retroviral envelope proteins mediate receptor recognition and membrane fusion during early infection. Endogenous envelope proteins may have kept, lost or modified their original function during evolution. This endogenous envelope protein has lost its fusogenic properties. It can inhibit cell growth through decrease expression of cyclin B1 and increased expression of p21 in vitro.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ERV3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ERV3-1
Gene Accession No. Q14264
Gene Alias endogenous retroviral sequence 3; endogenous retrovirus group 3 member 1; endogenous retrovirus group 3 member 1 Env polyprotein; endogenous retrovirus group 3, member 1; Envelope polyprotein; envR; ERV3; ERV3 envelope protein; ERV-3 envelope protein; ERV3-1; ERV3-1 envelope protein; ERVR; ERV-R; ERV-R envelope protein; HERVR; HERV-R; HERV-R envelope protein; HERV-R_7q21.2 provirus ancestral Env polyprotein; SU; Surface protein; TM; Transmembrane protein
Gene Symbols ERV3-1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ERV3-1. Recombinant protein control fragment (Product #RP-104560).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2086
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.